Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfaip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001137633.1 | NP_001131105.1 | 5 - 6 | 1365 | 122 | |
XM_006240577.2 | XP_006240639.1 | 4 - 5 | 1373 | 122 | ||
GenBank mRNA | FQ212109.1 | - | 4 - 5 | 1123 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |