Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025722.2 | NP_001020893.2 | 8 - 9 | 1130 | 61 | |
NM_001109936.1 | NP_001103406.1 | 9 - 10 | 679 | 61 | ||
NM_001109937.1 | NP_001103407.1 | 9 - 10 | 640 | 61 | ||
GenBank mRNA | BC098804.1 | - | 8 - 9 | 1130 | 7376321 | 61 |
DN932869.1 | - | 9 - 10 | 675 | 61 | ||
DN935585.1 | - | 9 - 10 | 699 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |