Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025766.1 | NP_001020937.1 | 1 - 2 | 403 | 111 | |
XM_006255599.2 | XP_006255661.1 | 2 - 3 | 766 | 111 | ||
XM_006255600.2 | XP_006255662.1 | 2 - 3 | 766 | 111 | ||
XM_008772527.1 | XP_008770749.1 | 2 - 3 | 471 | 111 | ||
GenBank mRNA | BC076391.1 | - | 2 - 3 | 529 | 111 | |
BC098808.1 | - | 1 - 2 | 403 | 7376793 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |