Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cirh1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009640.1 | NP_001009640.1 | 9 - 10 | 1129 | 109 | |
XM_006255562.1 | XP_006255624.1 | 9 - 10 | 1175 | 109 | ||
XM_006255563.2 | XP_006255625.1 | 9 - 10 | 1338 | 109 | ||
XM_008772488.1 | XP_008770710.1 | 9 - 10 | 1128 | 109 | ||
GenBank mRNA | BC088461.1 | - | 9 - 10 | 1129 | 7127580 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |