Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rwdd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_147146.2 | NP_671487.1 | 5 - 6 | 679 | 141 | |
XM_008772869.1 | XP_008771091.1 | 5 - 6 | 811 | 141 | ||
GenBank mRNA | FQ218614.1 | - | 2 - 3 | 214 | 141 | |
FQ220470.1 | - | 2 - 3 | 214 | 141 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |