Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022707.2 | NP_073198.1 | 1 - 2 | 72 | 59 | |
GenBank mRNA | CK356371.1 | - | 1 - 2 | 69 | 59 | |
FM050525.1 | - | 1 - 2 | 72 | 59 | ||
S95849.1 | - | 1 - 2 | 50 | 59 | ||
X71068.1 | - | 1 - 2 | 65 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Rattus norvegicus 1700 Inventoried Probe spans exons |