Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Khdrbs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133318.1 | NP_579852.1 | 4 - 5 | 632 | 79 | |
XM_008766952.1 | XP_008765174.1 | 4 - 5 | 652 | 79 | ||
GenBank mRNA | AF305618.1 | - | 4 - 5 | 632 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |