Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rsbn1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135872.1 | NP_001129344.1 | 6 - 7 | 2196 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |