Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gatsl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025128.1 | NP_001020299.1 | 1 - 2 | 171 | 77 | |
XM_006251335.2 | XP_006251397.1 | 1 - 2 | 239 | 77 | ||
XM_006251336.2 | XP_006251398.1 | 1 - 2 | 239 | 77 | ||
GenBank mRNA | BC091274.1 | - | 1 - 2 | 171 | 7383249 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |