Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amigo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178144.1 | NP_835280.1 | 1 | 553 | 87 | |
GenBank mRNA | AY237731.1 | - | 1 | 553 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Rattus norvegicus Both primers and probe map within a single exon |