Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp11c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006257596.2 | XP_006257658.1 | 25 - 26 | 2970 | 136 | |
XM_006257597.2 | XP_006257659.1 | 25 - 26 | 3983 | 136 | ||
XM_006257599.2 | XP_006257661.1 | 25 - 26 | 3983 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Transporter Rattus norvegicus Probe spans exons |