Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc27a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106439.1 | NP_001099909.1 | 8 - 9 | 1847 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons |