Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022393.2 | NP_071788.2 | 7 - 8 | 814 | 74 | |
XM_006246803.2 | XP_006246865.1 | 7 - 8 | 852 | 74 | ||
XM_008767867.1 | XP_008766089.1 | 7 - 8 | 852 | 74 | ||
XM_008767868.1 | XP_008766090.1 | 7 - 8 | 696 | 74 | ||
XM_008767869.1 | XP_008766091.1 | 7 - 8 | 693 | 74 | ||
GenBank mRNA | BC091151.1 | - | 7 - 8 | 814 | 7375234 | 74 |
FQ221204.1 | - | 7 - 8 | 703 | 74 | ||
FQ228437.1 | - | 7 - 8 | 672 | 74 | ||
J05495.1 | - | 7 - 8 | 674 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |