Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004272.1 | NP_001004272.1 | 2 - 3 | 215 | 93 | |
XM_006246747.2 | XP_006246809.1 | 2 - 3 | 271 | 93 | ||
XM_006246748.1 | XP_006246810.1 | 2 - 3 | 166 | 93 | ||
XM_008767840.1 | XP_008766062.1 | 2 - 3 | 166 | 93 | ||
GenBank mRNA | BC079162.1 | - | 2 - 3 | 215 | 7129493 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |