Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100756.1 | NP_001094226.1 | 14 - 15 | 1892 | 98 | |
XM_008764464.1 | XP_008762686.1 | 7 - 8 | 2961 | 98 | ||
GenBank mRNA | BC168226.1 | - | 14 - 15 | 1892 | 7371309 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |