Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100756.1 | NP_001094226.1 | 18 - 19 | 2404 | 64 | |
XM_008764464.1 | XP_008762686.1 | 11 - 12 | 3473 | 64 | ||
GenBank mRNA | BC098811.1 | - | 3 - 4 | 361 | 64 | |
BC168226.1 | - | 18 - 19 | 2404 | 7371309 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |