Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prr11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108287.1 | NP_001101757.1 | 7 - 8 | 936 | 74 | |
XM_008768073.1 | XP_008766295.1 | 7 - 8 | 1093 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |