Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dus4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135803.1 | NP_001129275.1 | 3 - 4 | 415 | 106 | |
XM_006239990.2 | XP_006240052.1 | 3 - 4 | 346 | 106 | ||
XM_006239991.1 | XP_006240053.1 | 4 - 5 | 573 | 106 | ||
XM_006239992.2 | XP_006240054.1 | 4 - 5 | 850 | 106 | ||
XM_006239993.2 | XP_006240055.1 | 2 - 3 | 259 | 106 | ||
XM_008764601.1 | XP_008762823.1 | 2 - 3 | 221 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |