Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akap17b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002730183.4 | XP_002730229.1 | 3 - 4 | 1142 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons |