Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E2f8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006229263.2 | XP_006229325.1 | 4 - 5 | 1277 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |