Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Peli3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127542.1 | NP_001121014.1 | 5 - 6 | 727 | 90 | |
GenBank mRNA | BC161922.1 | - | 5 - 6 | 713 | 8370772 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |