Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013032.3 | NP_037164.3 | 5 - 6 | 572 | 62 | |
GenBank mRNA | BC061743.1 | - | 5 - 6 | 572 | 5599027 | 62 |
D63772.1 | - | 603 | 62 | |||
U21104.1 | - | 5 - 6 | 584 | 62 | ||
U39555.1 | - | 5 - 6 | 662 | 62 | ||
X94255.1 | - | 5 - 6 | 629 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Rattus norvegicus Probe spans exons |