Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013151.2 | NP_037283.2 | 11 - 12 | 1325 | 121 | |
XM_006253347.2 | XP_006253409.1 | 11 - 12 | 1397 | 121 | ||
XM_008771348.1 | XP_008769570.1 | 12 - 13 | 1455 | 121 | ||
GenBank mRNA | BC061565.1 | - | 11 - 12 | 1325 | 6920438 | 121 |
M23697.1 | - | 10 - 11 | 1249 | 121 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |