Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nop56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025732.1 | NP_001020903.1 | 6 - 7 | 815 | 98 | |
GenBank mRNA | BC086568.1 | - | 3 - 4 | 493 | 98 | |
BC099149.1 | - | 6 - 7 | 815 | 7366316 | 98 | |
FQ232640.1 | - | 1 - 2 | 61 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |