Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif3g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013095.1 | NP_001013113.1 | 3 - 4 | 164 | 77 | |
GenBank mRNA | BC086383.1 | - | 3 - 4 | 164 | 7301572 | 77 |
FQ213005.1 | - | 3 - 4 | 205 | 77 | ||
FQ225883.1 | - | 3 - 4 | 77 | |||
FQ226276.1 | - | 3 - 4 | 196 | 77 | ||
FQ230888.1 | - | 3 - 4 | 190 | 77 | ||
FQ232595.1 | - | 3 - 4 | 194 | 77 | ||
FQ234727.1 | - | 1 - 2 | 46 | 77 | ||
FQ234921.1 | - | 3 - 4 | 195 | 77 | ||
FQ234929.1 | - | 3 - 4 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |