Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134530.1 | NP_001128002.1 | 4 - 5 | 658 | 86 | |
GenBank mRNA | BC169019.1 | - | 4 - 5 | 658 | 9091657 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Protease Mammalian Gene Collection Rattus norvegicus Inventoried Probe spans exons |