Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108754.1 | NP_001102224.1 | 7 - 8 | 862 | 95 | |
GenBank mRNA | FQ215152.1 | - | 7 - 8 | 728 | 95 | |
FQ226445.1 | - | 7 - 8 | 728 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |