Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ska3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108379.1 | NP_001101849.1 | 6 - 7 | 1222 | 108 | |
XM_006252074.1 | XP_006252136.1 | 6 - 7 | 1028 | 108 | ||
XM_006252075.1 | XP_006252137.1 | 6 - 7 | 1028 | 108 | ||
XM_008770758.1 | XP_008768980.1 | 6 - 7 | 1025 | 108 | ||
GenBank mRNA | BC166461.1 | - | 6 - 7 | 999 | 9030107 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |