Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Suco | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199403.1 | NP_955435.1 | 20 - 21 | 3029 | 89 | |
XM_006250164.2 | XP_006250226.1 | 20 - 21 | 3484 | 89 | ||
XM_006250166.2 | XP_006250228.1 | 19 - 20 | 3463 | 89 | ||
XM_008769657.1 | XP_008767879.1 | 20 - 21 | 3496 | 89 | ||
XM_008769658.1 | XP_008767880.1 | 19 - 20 | 3475 | 89 | ||
XM_008769659.1 | XP_008767881.1 | 20 - 21 | 3496 | 89 | ||
GenBank mRNA | AJ421447.1 | - | 20 - 21 | 3029 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus Probe spans exons |