Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025676.1 | NP_001020847.1 | 5 - 6 | 428 | 121 | |
GenBank mRNA | BC097991.1 | - | 5 - 6 | 428 | 7446014 | 121 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |