Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749167.3 | XP_003749215.1 | 3 - 4 | 266 | 94 | |
XM_006231788.2 | XP_006231850.1 | 3 - 4 | 266 | 94 | ||
XM_008760655.1 | XP_008758877.1 | 3 - 4 | 266 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |