Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fxr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100647.1 | NP_001094117.1 | 8 - 9 | 854 | 59 | |
GenBank mRNA | BC160929.1 | - | 8 - 9 | 854 | 8364923 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |