Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270954.1 | NP_001257883.1 | 1 - 2 | 381 | 59 | |
XM_008767871.1 | XP_008766093.1 | 2 - 3 | 2333 | 59 | ||
XM_008767872.1 | XP_008766094.1 | 2 - 3 | 2337 | 59 | ||
XM_008767873.1 | XP_008766095.1 | 3 - 4 | 4986 | 59 | ||
GenBank mRNA | AF239157.1 | - | 1 - 2 | 409 | 59 | |
BC099136.1 | - | 1 - 2 | 381 | 7455971 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |