Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006247233.2 | XP_006247295.1 | 2 - 3 | 399 | 72 | |
XM_006247234.2 | XP_006247296.1 | 2 - 3 | 403 | 72 | ||
XM_006247235.2 | XP_006247297.1 | 2 - 3 | 398 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons |