Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053786.1 | NP_446238.1 | 1 - 2 | 131 | 64 | |
GenBank mRNA | BC082110.1 | - | 1 - 2 | 131 | 7135071 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |