Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277194.1 | NP_001264123.1 | 5 - 6 | 813 | 145 | |
GenBank mRNA | FM122421.1 | - | 2 - 3 | 338 | 145 | |
FQ214725.1 | - | 5 - 6 | 794 | 145 | ||
FQ216469.1 | - | 5 - 6 | 797 | 145 | ||
FQ220496.1 | - | 5 - 6 | 790 | 145 | ||
FQ222191.1 | - | 5 - 6 | 795 | 145 | ||
FQ222396.1 | - | 5 - 6 | 795 | 145 | ||
FQ224965.1 | - | 5 - 6 | 792 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |