Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134865.1 | NP_001128337.1 | 9 - 10 | 2332 | 79 | |
XM_008765548.1 | XP_008763770.1 | 9 - 10 | 2354 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |