Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Caps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108990.1 | NP_001102460.1 | 2 - 3 | 168 | 135 | |
XM_006241337.2 | XP_006241399.1 | 12 - 13 | 1516 | 135 | ||
XM_008765359.1 | XP_008763581.1 | 11 - 12 | 1048 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |