Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bysl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182674.3 | NP_872615.3 | 6 - 7 | 1033 | 72 | |
GenBank mRNA | AY257675.4 | - | 6 - 7 | 1016 | 72 | |
BC079030.1 | - | 6 - 7 | 1022 | 7114086 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |