Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mocs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106881.1 | NP_001100351.1 | 5 - 6 | 725 | 62 | |
XM_006244415.2 | XP_006244477.1 | 6 - 7 | 795 | 62 | ||
XM_006244416.2 | XP_006244478.1 | 5 - 6 | 736 | 62 | ||
XM_006244418.2 | XP_006244480.1 | 6 - 7 | 790 | 62 | ||
XM_008766840.1 | XP_008765062.1 | 2 - 3 | 143 | 62 | ||
XM_008766841.1 | XP_008765063.1 | 5 - 6 | 734 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |