Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scn2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012647.1 | NP_036779.1 | 8 - 9 | 1242 | 65 | |
GenBank mRNA | M22254.1 | - | 8 - 9 | 1242 | 65 | |
X03639.1 | - | 8 - 9 | 1242 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Rattus norvegicus Probe spans exons |