Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100669.1 | NP_001094139.1 | 7 - 8 | 1047 | 59 | |
GenBank mRNA | BC161927.1 | - | 7 - 8 | 1041 | 8374170 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |