Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106752.1 | NP_001100222.1 | 8 - 9 | 918 | 60 | |
XM_006240422.2 | XP_006240484.1 | 10 - 11 | 1006 | 60 | ||
XM_006240423.2 | XP_006240485.1 | 9 - 10 | 975 | 60 | ||
XM_006240424.2 | XP_006240486.1 | 10 - 11 | 984 | 60 | ||
XM_008764762.1 | XP_008762984.1 | 8 - 9 | 764 | 60 | ||
GenBank mRNA | BC161880.1 | - | 9 - 10 | 950 | 7126409 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |