Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eno1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109908.1 | NP_001103378.1 | 2 - 3 | 255 | 95 | |
NM_012554.3 | NP_036686.2 | 2 - 3 | 206 | 95 | ||
XM_006239444.1 | XP_006239506.1 | 2 - 3 | 222 | 95 | ||
GenBank mRNA | BC063174.1 | - | 2 - 3 | 199 | 95 | |
BC078896.1 | - | 2 - 3 | 255 | 7112523 | 95 | |
BC081847.1 | - | 2 - 3 | 179 | 7189453 | 95 | |
BC090069.1 | - | 2 - 3 | 136 | 7108936 | 95 | |
BC091572.1 | - | 2 - 3 | 176 | 7107492 | 95 | |
BC097343.1 | - | 2 - 3 | 172 | 7461555 | 95 | |
CB809779.1 | - | 2 - 3 | 206 | 95 | ||
FQ214707.1 | - | 2 - 3 | 128 | 95 | ||
X02610.1 | - | 2 - 3 | 195 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |