Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adh6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106475.1 | NP_001099945.1 | 4 - 5 | 724 | 56 | |
XM_006233371.2 | XP_006233433.1 | 5 - 6 | 806 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |