Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrriq3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024307.1 | NP_001019478.1 | 5 - 6 | 1009 | 139 | |
GenBank mRNA | BC078994.1 | - | 5 - 6 | 1009 | 7112662 | 139 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |