Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Herpud2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024988.1 | NP_001020159.1 | 5 - 6 | 956 | 108 | |
XM_006242723.2 | XP_006242785.1 | 5 - 6 | 1134 | 108 | ||
XM_006242724.2 | XP_006242786.1 | 5 - 6 | 1196 | 108 | ||
GenBank mRNA | BC081861.1 | - | 5 - 6 | 956 | 7134757 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |