Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plin5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134637.1 | NP_001128109.1 | 2 - 3 | 160 | 96 | |
XM_008766770.1 | XP_008764992.1 | 2 - 3 | 203 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |