Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Otud5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037496.1 | NP_001032585.1 | 5 - 6 | 884 | 92 | |
XM_006256737.2 | XP_006256799.1 | 5 - 6 | 1534 | 92 | ||
XM_006256738.2 | XP_006256800.1 | 5 - 6 | 1533 | 92 | ||
XM_006256739.2 | XP_006256801.1 | 5 - 6 | 1532 | 92 | ||
XM_006256740.2 | XP_006256802.1 | 6 - 7 | 1404 | 92 | ||
XM_006256741.2 | XP_006256803.1 | 6 - 7 | 1403 | 92 | ||
GenBank mRNA | BC110054.1 | - | 5 - 6 | 884 | 7446041 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |