Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009176.2 | NP_001009176.1 | 1 - 2 | 784 | 98 | |
GenBank mRNA | BC129078.1 | - | 1 - 2 | 784 | 8371876 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |